Order Entry
Czech Republic
ContactUsLinkComponent
5-(Ethylthio)-1H-tetrazole (ETT) 0.5M in acetonitrile, BioSyn™ activator reagent, for DNA/RNA synthesis, Burdick & Jackson™
Irritant
Health
Flammable
5-(Ethylthio)-1H-tetrazole (ETT) 0.5M in acetonitrile, BioSyn™ activator reagent, for DNA/RNA synthesis, Burdick & Jackson™
Certificates
Irritant
Health
Flammable
5-(Ethylthio)-1H-tetrazole (ETT) 0.5M in acetonitrile, BioSyn™ activator reagent, for DNA/RNA synthesis, Burdick & Jackson™

Specification Test Results

  • Appearance:
    complying
  • Identity (IR):
    complying
  • Assay Of 5-(Ethylthio)-1H-Tetrazole (Ett):
    0.48 - 0.52 mol/l
  • Water (Karl Fischer):
    Max. 60 ppm

Specification Test Results

Frequently Bought Together