Order Entry
Czech Republic
ContactUsLinkComponent
Sodium hydroxide 0.1 mol/l Reag. Ph.Eur. volumetric solution, Burdick & Jackson™
Sodium hydroxide 0.1 mol/l Reag. Ph.Eur. volumetric solution, Burdick & Jackson™
Catalog # HONC65879-10L
CAS Number:  
Sodium hydroxide 0.1 mol/l Reag. Ph.Eur. volumetric solution, Burdick & Jackson™
Catalog # HONC65879-10L
CAS Number:  

Specifications

  • Pack type:
    Fiberboard box
  • Pk:
    10 L
  • Seq:
    5214

Specifications

About this item

 :
 :40 g/mol
Boiling Pt:100 °C (1.013 hPa)
Density:1.000 g/cm3 (20 °C)
Storage Temperature:Ambient
UN:1824
ADR:8,III
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548