Order Entry
Czech Republic
ContactUsLinkComponent
L(+)-Potassium sodium tartrate tetrahydrate ≥99.5% (by perchloric acid titration), Puriss. p.a.
L(+)-Potassium sodium tartrate tetrahydrate ≥99.5% (by perchloric acid titration), Puriss. p.a.
Catalog # HONC32339H-25KG
CAS Number:  
L(+)-Potassium sodium tartrate tetrahydrate ≥99.5% (by perchloric acid titration), Puriss. p.a.
Catalog # HONC32339H-25KG
CAS Number:  

Specifications

  • Pack type:
    Paper/plastic bag
  • Pk:
    25 kg
  • Seq:
    4283

Specifications

Specification Test Results

  • Assay (titration with Perchloric acid):
    min. 99.5 %
  • pH (5 %, 20°C):
    7.0 - 8.5
  • Iron (Fe):
    max. 0.0005 %
  • Heavy metals (as Pb):
    max. 0.0005 %

Specification Test Results

About this item

 :
 :282.22 g/mol
Melting Pt:70 - 80 °C
Melting Pt:70…80 °C
Density:1.79 g/cm3 (20 °C)
Storage Temperature:Ambient
EINECS:206-156-8
CAS Number:6381-59-5
Formula:KOOC(CHOH)₂COONa·4H₂O
MDL Number:MFCD00150989