1,2-Propanediol Analytical standard, reference material, Fluka™
HONCR1921-1G
:
- Pack type:Glass Bottle
- Pk:1 g
- Standards:Analytical standard
- CAS No.:57-55-6
- Description:1,2-Propanediol
- Storage conditions:RT
- MDL No.:MFCD00064272
- Synonym(s):1,2-Propylene glycol
- Melting point:-60 °C
- Formula:C₃H₈O₂
- Molecular weight:76.09 g/mol
- Description:1,2-Propanediol
- Flash point:103 °C
- Physical form:Liquid
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery