sulphamethazine Analytical standard, reference material, Fluka™
HONCR1883-100MG
:
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:57-68-1
- Description:Sulphamethazine
- Storage conditions:RT
- MDL No.:MFCD00006066
- Synonym(s):4-Amino-N-(4,6-dimethyl-2-pyrimidinyl)benzenesulfonamide
- Melting point:197 °C
- Formula:C₁₂H₁₄N₄O₂S
- Molecular weight:278.33 g/mol
- Description:Sulphamethazine
- Physical form:powder
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery