VWR®, Perfluorohexane-1-sulphonic acid (PFHxS), 2 µg/ml in MeOH, Certified Reference Material (CRM)
Catalog # 91407.010
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass ampoule
- Pk:1 mL
- Standards:CRM ISO 17025/17034
- CAS No.:355-46-4
- Description:Perfluorohexane-1-sulphonic acid (PFHxS), 2 µg/ml in MeOH
- UN No.:1230
- Synonym(s):PFHxS
- Concentration:2 µg/ml in MeOH
- Description:Perfluorohexane-1-sulphonic acid
Specifications
About this item
Per- and polyfluoroalkyl substances (PFAS) are amongst the primary emerging contaminants of concern. The detection and quantification of known PFAS and the discovery of unknown PFAS substances has never been more important.
VWR® is supplying a comprehensive range of standards and CRMs to enable this environmental analysis.
Certifications: Certified Reference Material (CRM) ISO 17025 / ISO 17034