VWR®, Perfluoropentanoic acid, Certified Reference Material (CRM)
Catalog # 91355.001
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass vial
- Pk:100 mg
- Standards:CRM ISO 17025/17034
- CAS No.:2706-90-3
- Description:Perfluoropentanoic acid
- UN No.:3265
- Synonym(s):Per- and polyfluoroalkyl substances (PFAS)
- Formula:C₅HF₉O₂
- Molecular weight:264,05 g/mol
- Description:Perfluoropentanoic acid
Specifications
About this item
Per- and polyfluoroalkyl substances (PFAS) are amongst the primary emerging contaminants of concern. The detection and quantification of known PFAS and the discovery of unknown PFAS substances has never been more important.
VWR® is supplying a comprehensive range of standards and CRMs to enable this environmental analysis.
Certifications: Certified Reference Material (CRM) ISO 17025 / ISO 17034