Order Entry
Czech Republic
ContactUsLinkComponent
1-(tert-Butoxycarbonyl)-2-pyrrolidinone 97%
Irritant
1-(tert-Butoxycarbonyl)-2-pyrrolidinone 97%
Certificates
Irritant
1-(tert-Butoxycarbonyl)-2-pyrrolidinone 97%

Specification Test Results

  • Appearance (Color):
    Clear colorless to yellow to brown
  • Appearance (Form):
    Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4645 to 1.4670 (20°C, 589 nm)
  • GC:
    ≥96.0 %

Specification Test Results

Frequently Bought Together