Glass screw top 0,3 ml microvials for <2 ml samples, level 3 high performance applications, SureSTART™
Supplier: Thermo Fisher Scientific
Certificates
About this item
Choose Thermo Scientific™ SureSTART™ 0,3 ml glass screw top microvials, performance level 3, when you need to maximise the injection volume for <2 ml samples.
This 12×32 mm microvials has a fixed insert that provides a total usable volume of 0,25 ml and a residual volume of 4 µl. Available in clear and amber glass, this is a performance level 3 vial, which means it is designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyses.
Limited Time Sale
Select items below are currently on sale
Caps
Product Information
Frequently Bought Together