
2,2,6,6-Tetramethylpiperidinooxy (TEMPO) (free radical) ≥98% for synthesis
Catalog # 91304.100
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Assay:Min 98%
Specification Test Results
About this item
:
:156.25 g/mol
Storage Temperature:Ambient
EINECS:219-888-8
UN:3263
ADR:8,II
:GHS05
CAS Number:2564-83-2
Formula:C₉H₁₈NO
MDL Number:MFCD00009599