Order Entry
Czech Republic
ContactUsLinkComponent
 
GemCell™ U.S. Human Serum AB
  100-512-100
undefined
GemCell™ U.S. Human Serum AB
  100-512-100

 

  • Description:
    U.S. human serum AB
  • Seq:
    01
  • Origin:
    US
  • Storage temperature:
    Store frozen at −20…−10 °C
  • Pack type:
    Bottle
  • Pk:
    100 mL

 

 

GemCell™ human serum AB is collected from healthy male donors of the AB serotype at FDA-licensed facilities in the United States. This material is defibrinated from source plasma AB. All donor units are tested for viral markers and found to be non-reactive.

  • 0,1 µm sterile-filtered
  • Gamma irradiation available