Amlodipine Besylate, Reference Standard
1029501.
: USP
:
- Pack type:Vial
- Pk:350 mg
- Standards:USP reference standard
- CAS No.:111470-99-6
- Description:Amlodipine Besylate
- Seq:01
- UN No.:3077
- Formula:C₂₆H₃₁ClN₂O₈S
- Description:Amlodipine Besylate
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard