Order Entry
Czech Republic
ContactUsLinkComponent
 
Adapalene Related Compounds, Reference Standard
 :  USP
 
Adapalene Related Compounds, Reference Standard
 :  USP

 

  • CAS No.:
    932033-58-4
  • Synonym(s):
    2,2'-binaphthyl-6,6'-dicarboxylic acid
  • Formula:
    C₂₂H₁₄O₄
  • Description:
    Adapalene Related Compound E

 

Frequently Bought Together