Order Entry
Czech Republic
ContactUsLinkComponent
 
Acetaminophen Related Compounds, Reference Standard
 :  USP
 
Acetaminophen Related Compounds, Reference Standard
 :  USP

 

  • CAS No.:
    2623-33-8
  • Synonym(s):
    4-(acetylamino)phenyl acetate
  • Formula:
    C₁₀H₁₁NO₃
  • Description:
    Acetaminophen Related Compound A

 

Frequently Bought Together