Order Entry
Czech Republic
ContactUsLinkComponent
 
16alpha-Hydroxyprednisolone, Pharmaceutical Analytical Impurities
  1A00880.
 :  USP
 :  
16alpha-Hydroxyprednisolone, Pharmaceutical Analytical Impurities
  1A00880.
 :  USP
 :  

 

  • Pack type:
    Vial
  • Pk:
    25 mg
  • Standards:
    Pharmaceutical Analytical Impurities
  • CAS No.:
    13951-70-7
  • Description:
    16alpha-Hydroxyprednisolone
  • Synonym(s):
    (8S,9S,10R,11S,13S,14S,16R,17S)-11,16,17-trihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-3H-cyclopenta[a]phenanthren-3-one
  • Description:
    16alpha-Hydroxyprednisolone

 

 

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.