16Alpha-Hydroxy-11-keto Prednisolone, Pharmaceutical Analytical Impurities
Catalog # 1A01250.
Supplier: USP
CAS Number:
Specifications
- Typ balení:Vial
- Bal.:25 mg
- Standard:Pharmaceutical Analytical Impurities
- Čí. CAS:3754-05-0
- Popis:16Alpha-Hydroxy-11-keto Prednisolone
- Synonym(s):8S,9S,10R,13S,14S,16R,17S)-16,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-7,8,9,10,12,13,14,15,16,17-decahydro-3H-cyclopenta[a]phenanthrene-3,11(6H)-dione
- Popis:16Alpha-Hydroxy-11-keto Prednisolone
Specifications
About this item
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.