Trandolapril Related Compounds, Reference Standard
Dodavatel: USP
Certifikáty
O tomto produktu
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifikace
- Synonym(s):(2S, 3aR, 7aS)-1-[N-[(S)-1-Carboxy-3-cyclohexylpropyl]-L-alanyl]hexahydro-2-indolinecarboxylic acid 1-ethyl ester hydrochloride
- Description:Trandolapril Related Compound C
- CAS No.:149881-40-3
Specifikace
Frequently Bought Together