Order Entry
Czech Republic
ContactUsLinkComponent
Sulfamethoxazole Related Compounds, Reference Standard
Sulfamethoxazole Related Compounds, Reference Standard
Supplier:  USP
Certificates
Sulfamethoxazole Related Compounds, Reference Standard
Supplier:  USP
Certificates

Specifications

  • CAS No.:
    1072-67-9
  • Synonym(s):
    5-methylisoxazol-3-amine
  • Formula:
    C₄H₆N₂O
  • Description:
    Sulfamethoxazole Related Compound C

Specifications

Related Information

Frequently Bought Together