Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
 

Rivastigmine Related Compounds, Reference Standard

 :   USP
Rivastigmine Related Compounds, Reference Standard
 :  USP
 
Rivastigmine Related Compounds, Reference Standard

 

 

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

 

 

 

  • Catalog No:
  • 1604811.
  • 1604869.
  • 1604870.
  • 1604950.
  • 1604961.
  • 1604825.
  • Description:
  • Rivastigmine Related Compound C
  • Rivastigmine Related Compound A
  • Rivastigmine Related Compound B
  • Rivastigmine Related Compound E
  • Rivastigmine Related Compound F
  • Rivastigmine Related Compound D
  • Synonym(s):
  • (S)-3-[1-(Dimethylamino)ethyl]phenol
  • (+)-Di-(p-toluoyl)-D-tartaric acid
  • (R,S)-3-[1-(Dimethylamino)ethyl]phenyl dimethylcarbamate
  • (S)-1-(3-{[Ethyl(methyl)carbamoyl]oxy}phenyl)-N,N,N-trimethylethanaminium chloride
  • 3-Vinylphenyl ethyl(methyl)carbamate
  • 3-Acetylphenyl ethyl(methyl)carbamate
  • CAS No.:
  • 139306-10-8
  • 32634-68-7
  • 25081-93-0
  • 1346602-84-3
  • 855300-09-3
  • Formula:
  • C₁₀H₁₅NO
  • C₂₀H₁₈O₈
  • C₁₃H₂₀N₂O₂
  • C₁₂H₁₅NO₂
  • C₁₂H₁₅NO₃
  • UN No.:
  • 2810
  • 2811

 

Product Family Options

 

  • Description
    Standards
    Pack type
    Pk
     
     
  • Rivastigmine Related Compound C
    USP reference standard
    Vial
    15 mg
       
       
    1 * 15 mg
    1 * 15 mg - 26 990,00 Kč
    26 990,00 Kč
    1 * 15 mg
       26 990,00 Kč

     
     
    Specifications
    Cat. No.
    1604811.
    Description
    Rivastigmine Related Compound C
    Synonym(s)
    (S)-3-[1-(Dimethylamino)ethyl]phenol
    CAS No.
    139306-10-8
    Formula
    C₁₀H₁₅NO
    UN No.
  • Rivastigmine Related Compound A
    USP reference standard
    Vial
    15 mg
       
       
    1 * 15 mg
    1 * 15 mg - 25 820,00 Kč
    25 820,00 Kč
    1 * 15 mg
       25 820,00 Kč

     
     
    Specifications
    Cat. No.
    1604869.
    Description
    Rivastigmine Related Compound A
    Synonym(s)
    (+)-Di-(p-toluoyl)-D-tartaric acid
    CAS No.
    32634-68-7
    Formula
    C₂₀H₁₈O₈
    UN No.
  • Rivastigmine Related Compound B
    USP reference standard
    Ampule
    15 mg
       
       
    1 * 15 mg
    1 * 15 mg - 27 110,00 Kč
    27 110,00 Kč
    1 * 15 mg
       27 110,00 Kč

     
     
    Specifications
    Cat. No.
    1604870.
    Description
    Rivastigmine Related Compound B
    Synonym(s)
    (R,S)-3-[1-(Dimethylamino)ethyl]phenyl dimethylcarbamate
    CAS No.
    25081-93-0
    Formula
    C₁₃H₂₀N₂O₂
    UN No.
    2810
  • Rivastigmine Related Compound E
    USP reference standard
    Vial
    15 mg
       
       
    1 * 15 mg
    1 * 15 mg - 29 200,00 Kč
    29 200,00 Kč
    1 * 15 mg
       29 200,00 Kč

     
     
    Specifications
    Cat. No.
    1604950.
    Description
    Rivastigmine Related Compound E
    Synonym(s)
    (S)-1-(3-{[Ethyl(methyl)carbamoyl]oxy}phenyl)-N,N,N-trimethylethanaminium chloride
    CAS No.
    Formula
    UN No.
    2811
  • Rivastigmine Related Compound F
    USP reference standard
    Ampule
    25 mg
       
       
    1 * 25 mg
    1 * 25 mg - 28 650,00 Kč
    28 650,00 Kč
    1 * 25 mg
       28 650,00 Kč

     
     
    Specifications
    Cat. No.
    1604961.
    Description
    Rivastigmine Related Compound F
    Synonym(s)
    3-Vinylphenyl ethyl(methyl)carbamate
    CAS No.
    1346602-84-3
    Formula
    C₁₂H₁₅NO₂
    UN No.
  • Rivastigmine Related Compound D
    USP reference standard
    Vial
    50 mg
       
       
    1 * 50 mg
    1 * 50 mg - 25 050,00 Kč
    25 050,00 Kč
    1 * 50 mg
       25 050,00 Kč

     
     
    Specifications
    Cat. No.
    1604825.
    Description
    Rivastigmine Related Compound D
    Synonym(s)
    3-Acetylphenyl ethyl(methyl)carbamate
    CAS No.
    855300-09-3
    Formula
    C₁₂H₁₅NO₃
    UN No.