

Methanol with 0,1% (v/v) formic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
:64-18-6
:64.7 °C (main component)
:–97.68 °C (main component)
:0.792 g/cm³ (20 °C) (main component)
:11 °C
:1230
:3,II
:GHS02
:GHS08
:Ambient
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Assay (GC):Min. 99.5 %
- Calcium (Ca):Max. 0.5 ppm
- Potassium (K):Max. 0.5 ppm
- Magnesium (Mg):Max. 0.5 ppm
- Sodium (Na):Max. 2 ppm
- Transmittance at 254 nm:Min. 90 %
- Transmittance at 230 nm:Min. 10 %
- HPLC Gradient at 254 nm:Max. 10 mAU
- LC-MS application:complying
Frequently Bought Together