Order Entry
Czech Republic
ContactUsLinkComponent
Pemetrexed Disodium Enantiomer, Pharmaceutical Analytical Impurities
Pemetrexed Disodium Enantiomer, Pharmaceutical Analytical Impurities
Catalog # 1A00620.
Supplier:  USP
CAS Number:  
Pemetrexed Disodium Enantiomer, Pharmaceutical Analytical Impurities
Catalog # 1A00620.
Supplier:  USP
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    25 mg
  • Standards:
    Pharmaceutical Analytical Impurities
  • CAS No.:
    937370-10-0
  • Description:
    Pemetrexed Disodium Enantiomer
  • Synonym(s):
    (2R)-2-[[4-[2-(2-amino-4-oxo-4,7-dihydro-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]pentanedioic acid, sodium salt
  • Description:
    Pemetrexed Disodium Enantiomer

Specifications

About this item

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.