Order Entry
Czech Republic
ContactUsLinkComponent
Orlistat Related Compounds, Reference Standard
Orlistat Related Compounds, Reference Standard
Supplier:  USP
Certificates
Orlistat Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    130793-27-0
  • UN No.:
    1648
  • Synonym(s):
    (3S,4R,6S)-3-Hexyl-2-oxo-6-undecyltetrahydro-2H-pyran-4-yl formyl-L-leucinate in acetonitrile
  • Description:
    Orlistat Solution Related Compound D
  • Formula:
    C₂₉H₅₃NO₅

Specifications

Frequently Bought Together