Order Entry
Czech Republic
ContactUsLinkComponent
Omeprazole Related Compounds, Reference Standard
Omeprazole Related Compounds, Reference Standard
Supplier:  USP
Certificates
Omeprazole Related Compounds, Reference Standard
Supplier:  USP
Certificates

Specifications

  • CAS No.:
    88546-55-8
  • Synonym(s):
    5-methoxy-2-[[(4-methoxy-3,5-dimethylpyridin-2-yl)methyl]sulfonyl]-1H-benzimidazole
  • Formula:
    C₁₇H₁₉N₃O₄S
  • Description:
    Omeprazole Related Compound A

Specifications

Related Information

Frequently Bought Together