Order Entry
Czech Republic
ContactUsLinkComponent
Nevirapine Related Compounds, Reference Standard
Nevirapine Related Compounds, Reference Standard
Supplier:  USP
Certificates
Nevirapine Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • Synonym(s):
    5,11-Dihydro-6H-11-propyl-4-methyl-dipyrido[3,2-b:2',3'-e] [1,4]diazepin-6-one
  • Description:
    Nevirapine Related Compound C
  • CAS No.:
    133627-17-5
  • UN No.:
    2811
  • Formula:
    C₁₄H₁₄N₄O

Specifications

Frequently Bought Together