Efavirenz Related Compounds, Reference Standard
Supplier: USP
Certificates
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications
- CAS No.:391860-73-4
- Synonym(s):6-chloro-2-cyclopropyl-4-(trifluoromethyl)quinoline
- Description:Efavirenz Related Compound C
Specifications
Frequently Bought Together