Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Choline Fenofibrate, Reference Standard

Choline Fenofibrate, Reference Standard

Choline Fenofibrate, USP reference standard, Vial, 50 mg
Catalog # 1133558.
Supplier:   USP
Choline Fenofibrate, Reference Standard
Catalog # 1133558.
Supplier:  USP
CAS Number:  
Choline Fenofibrate, Reference Standard

Choline Fenofibrate, USP reference standard, Vial, 50 mg

CAS Number:  
Price
14 080,00 Kč
Log in for availability
1 * 50 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    50 mg
  • Standards:
    USP reference standard
  • Description:
    Choline Fenofibrate
  • Cat. No.:
    1133558.
  • Description:
    Choline Fenofibrate
  • Synonym(s):
    2-Hydroxy-N,N,N-trimethylethanaminium 2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoate
  • CAS No.:
    856676-23-8

Specifications