Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Vigabatrin, Reference Standards

Vigabatrin, Reference Standards

Supplier:   USP
Vigabatrin, Reference Standards
Supplier:  USP
Certificates

4 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1712001.
  • 1712012.
  • 1712023.
  • 1712045.
  • Description:
  • Vigabatrin
  • Vigabatrin Related Compound A
  • Vigabatrin Related Compound B
  • Vigabatrin Related Compound E
  • Synonym(s):
  • 5-vinylpyrrolidin-2-one
  • (E)-2-(2-aminoethyl)but-2-enoic acid hydrochloride
  • 2-(2-aminobut-3-enyl)malonic acid
  • CAS No.:
  • 68506-86-5
  • 7529-16-0
  • UN No.:
  • 2810

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1712001.
    Vigabatrin
    Vigabatrin
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 13 060,00 Kč
    13 060,00 Kč
    1 * 200 mg
       13 060,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    1712001.
    Description
    Vigabatrin
    Synonym(s)
    CAS No.
    68506-86-5
    UN No.
  • 1712012.
    Vigabatrin Related Compound A
    Vigabatrin Related Compound A
    USP reference standard
    Ampule
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 28 370,00 Kč
    28 370,00 Kč
    1 * 50 mg
       28 370,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    1712012.
    Description
    Vigabatrin Related Compound A
    Synonym(s)
    5-vinylpyrrolidin-2-one
    CAS No.
    7529-16-0
    UN No.
    2810
  • 1712023.
    Vigabatrin Related Compound B
    Vigabatrin Related Compound B
    USP reference standard
    Vial
    30 mg
    Log in for availability
    Log in for availability
    1 * 30 mg
    1 * 30 mg - 30 400,00 Kč
    30 400,00 Kč
    1 * 30 mg
       30 400,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    1712023.
    Description
    Vigabatrin Related Compound B
    Synonym(s)
    (E)-2-(2-aminoethyl)but-2-enoic acid hydrochloride
    CAS No.
    UN No.
  • 1712045.
    Vigabatrin Related Compound E
    Vigabatrin Related Compound E
    USP reference standard
    Vial
    30 mg
    Log in for availability
    Log in for availability
    1 * 30 mg
    1 * 30 mg - 30 400,00 Kč
    30 400,00 Kč
    1 * 30 mg
       30 400,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    1712045.
    Description
    Vigabatrin Related Compound E
    Synonym(s)
    2-(2-aminobut-3-enyl)malonic acid
    CAS No.
    UN No.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.