Order Entry
Czech Republic
ContactUsLinkComponent
Triclabendazole Related Compounds, Reference Standard
Triclabendazole Related Compounds, Reference Standard
Supplier:  USP
Certificates
Triclabendazole Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    100648-13-3
  • Synonym(s):
    5-Chloro-6-(2,3-dichlorophenoxy)-2-(methylsulFood Ingredientsnyl)-1H-benzimidazole
  • Description:
    Triclabendazole Related Compound A

Specifications

Frequently Bought Together