Sulindac, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
4 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1642008.
- 1642019.
- 1642020.
- 1642031.
- Description:
- Sulindac
- Sulindac Related Compound A
- Sulindac Related Compound B
- Sulindac Related Compound C
- Synonym(s):
- (E)-2-{5-Fluoro-2-methyl-1-[4-(methylsulFood Ingredientsnyl)benzylidene]-1H-inden-3-yl}acetic acid
- (Z)-2-{5-fluoro-2-methyl-1-[4-(methylsulfonyl)benzylidene]-1H-inden-3-yl}acetic acid
- (Z)-2-{5-fluoro-2-methyl-1-[4-(methylthio)benzylidene]-1H-inden-3-yl}acetic acid
- CAS No.:
- 38194-50-2
- 53933-60-1
- 59973-80-7
- 49627-27-2
- UN No.:
- 2811
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1642008.SulindacSpecifications:
More
Specifications
Cat. No.1642008.DescriptionSulindacSynonym(s)CAS No.38194-50-2UN No.2811 - 1642019.Sulindac Related Compound ASulindac Related Compound AUSP reference standardVial20 mgLog in for availability1 * 20 mg30 680,00 KčSpecifications:
More
Specifications
Cat. No.1642019.DescriptionSulindac Related Compound ASynonym(s)(E)-2-{5-Fluoro-2-methyl-1-[4-(methylsulFood Ingredientsnyl)benzylidene]-1H-inden-3-yl}acetic acidCAS No.53933-60-1UN No. - 1642020.Sulindac Related Compound BSulindac Related Compound BUSP reference standardVial20 mgLog in for availability1 * 20 mg27 820,00 KčSpecifications:
More
Specifications
Cat. No.1642020.DescriptionSulindac Related Compound BSynonym(s)(Z)-2-{5-fluoro-2-methyl-1-[4-(methylsulfonyl)benzylidene]-1H-inden-3-yl}acetic acidCAS No.59973-80-7UN No. - 1642031.Sulindac Related Compound CSulindac Related Compound CUSP reference standardVial20 mgLog in for availability1 * 20 mg27 820,00 KčSpecifications:
More
Specifications
Cat. No.1642031.DescriptionSulindac Related Compound CSynonym(s)(Z)-2-{5-fluoro-2-methyl-1-[4-(methylthio)benzylidene]-1H-inden-3-yl}acetic acidCAS No.49627-27-2UN No.



