Order Entry
Czech Republic
ContactUsLinkComponent
Salicylic Acid, Reference Standards
Salicylic Acid, Reference Standards
Supplier:  USP
Certificates
undefined
Salicylic Acid, Reference Standards
Supplier:  USP
Certificates

Specifications

  • CAS No.:
    99-96-7
  • Synonym(s):
    4-hydroxybenzoic acid
  • Formula:
    C₇H₆O₃
  • Description:
    Salicylic Acid Related Compound A

Specifications

Related Information

Frequently Bought Together