Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Rotigotine, Reference Standards

Rotigotine, Reference Standards

Dodavatel:   USP
Rotigotine, Reference Standards
Dodavatel:  USP
Certifikáty

2 Dostupných variant níže

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifikace

Specifikace

  • Catalog No:
  • 1606060.
  • 1606081.
  • Description:
  • Rotigotine
  • Rotigotine Racemate
  • Synonym(s):
  • (S)-6-{Propyl[2-(thiophen-2-yl)ethyl]amino}-5,6,7,8-tetrahydronaphthalen-1-ol
  • (RS)-6-{Propyl[2-(thiophen-2-yl)ethyl]amino}-5,6,7,8-tetrahydronaphthalen-1-ol hydrochloride
  • CAS No.:
  • 99755-59-6
  • 102120-99-0
  • UN No.:
  • 3077
  • 3077

Specifikace

Product Family Options

Informace o produktu

  • Description
    Standards
    Pack type
    Pk
    Dostupnost
    Price
  • Rotigotine
    USP reference standard
    Vial
    50 mg
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 50 mg
    1 * 50 mg - 9 120,00 Kč
    9 120,00 Kč
    1 * 50 mg
       9 120,00 Kč

    Specifikace:
    More
    Specifications
    Cat. No.
    1606060.
    Description
    Rotigotine
    Synonym(s)
    (S)-6-{Propyl[2-(thiophen-2-yl)ethyl]amino}-5,6,7,8-tetrahydronaphthalen-1-ol
    CAS No.
    99755-59-6
    UN No.
    3077
  • Rotigotine Racemate
    USP reference standard
    Vial
    25 mg
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 25 mg
    1 * 25 mg - 19 750,00 Kč
    19 750,00 Kč
    1 * 25 mg
       19 750,00 Kč

    Specifikace:
    More
    Specifications
    Cat. No.
    1606081.
    Description
    Rotigotine Racemate
    Synonym(s)
    (RS)-6-{Propyl[2-(thiophen-2-yl)ethyl]amino}-5,6,7,8-tetrahydronaphthalen-1-ol hydrochloride
    CAS No.
    102120-99-0
    UN No.
    3077
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.