Order Entry
Czech Republic
ContactUsLinkComponent
Rosuvastatin Lactone, Pharmaceutical Analytical Impurities
Rosuvastatin Lactone, Pharmaceutical Analytical Impurities
Catalog # 1A01200.
Supplier:  USP
CAS Number:  
Rosuvastatin Lactone, Pharmaceutical Analytical Impurities
Catalog # 1A01200.
Supplier:  USP
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    25 mg
  • Standards:
    Pharmaceutical Analytical Impurities
  • CAS No.:
    503610-43-3
  • Description:
    Rosuvastatin Lactone
  • Synonym(s):
    N-[4-(4-Fluorophenyl)-5-{(E)-2-[(2S,4R)-4-hydroxy-6-oxotetrahydro-2H-pyran-2-yl]vinyl}-6-isopropylpyrimidin-2-yl]-N-methylmethanesulfonamide
  • Description:
    Rosuvastatin Lactone

Specifications

About this item

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.