Rivaroxaban Open Chain Dithiophene Analog, Pharmaceutical Analytical Impurities
Catalog # 1A01460.
Dodavatel: USP
Číslo CAS:
Specifikace
- Pack type:Vial
- Pk:10 mg
- Standards:Pharmaceutical Analytical Impurities
- CAS No.:1807455-76-0
- Description:Rivaroxaban Open Chain Dithiophene Analog
- Synonym(s):(S)-5-chloro-N-(4-(5-((5-chlorothiophene-2-carboxamido)methyl)-2-oxooxazolidin-3-yl)phenyl)-N-(2-(2-(methylamino)-2-oxoethoxy)ethyl)thiophene-2-carboxamide
- Description:Rivaroxaban Open Chain Dithiophene Analog
Specifikace
O tomto produktu
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.