Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
 

Regorafenib, Reference Standards

 :   USP
Regorafenib, Reference Standards
 :  USP
 
Regorafenib, Reference Standards

 

 

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

 

 

 

  • Catalog No:
  • 1600201.
  • 1600212.
  • Description:
  • Regorafenib
  • Regorafenib Related Compound A
  • Synonym(s):
  • 4-(4-Amino-3-fluorophenoxy)-N-methylpicolinamide
  • CAS No.:
  • 1019206-88-2
  • 757251-39-1

 

Product Family Options

 

  • Description
    Standards
    Pack type
    Pk
     
     
  • Regorafenib
    USP reference standard
    Vial
    200 mg
       
       
    1 * 200 mg
    1 * 200 mg - 11 590,00 Kč
    11 590,00 Kč
    1 * 200 mg
       11 590,00 Kč

     
     
    Specifications
    Cat. No.
    1600201.
    Description
    Regorafenib
    Synonym(s)
    CAS No.
    1019206-88-2
  • Regorafenib Related Compound A
    USP reference standard
    Vial
    15 mg
       
       
    1 * 15 mg
    1 * 15 mg - 22 640,00 Kč
    22 640,00 Kč
    1 * 15 mg
       22 640,00 Kč

     
     
    Specifications
    Cat. No.
    1600212.
    Description
    Regorafenib Related Compound A
    Synonym(s)
    4-(4-Amino-3-fluorophenoxy)-N-methylpicolinamide
    CAS No.
    757251-39-1