Pemetrexed, Pharmaceutical Analytical Impurities
Catalog # 1A01140.
Supplier: USP
CAS Number:
Specifications
- Pack type:Vial
- Pk:25 mg
- Standards:Pharmaceutical Analytical Impurities
- CAS No.:137281-39-1
- Description:Pemetrexed Acid
- Synonym(s):4-(2-(2-amino-4-oxo-4,7-dihydro-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl)benzoic acid
- Description:Pemetrexed Acid
Specifications
About this item
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.