Order Entry
Czech Republic
ContactUsLinkComponent
 
Isobutylglutarmonoamide, Pharmaceutical Analytical Impurities
Catalog # 1A01840.
Supplier:  USP
CAS Number:  
Isobutylglutarmonoamide, Pharmaceutical Analytical Impurities
Catalog # 1A01840.
Supplier:  USP
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    25 mg
  • Standards:
    Pharmaceutical Analytical Impurities
  • CAS No.:
    181289-15-6
  • Description:
    Isobutylglutarmonoamide
  • Synonym(s):
    3-(2-Amino-2-oxoethyl)-5-methylhexanoic Acid
  • Description:
    Isobutylglutarmonoamide

Specifications

About this item

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.