Diphthalimido Morpholinone Analog, Pharmaceutical Analytical Impurities
Catalog # 1A01600.
Supplier: USP
CAS Number:
Specifications
- Pack type:Vial
- Pk:25 mg
- Standards:Pharmaceutical Analytical Impurities
- CAS No.:1643354-27-1
- Description:Diphthalimido Morpholinone Analog
- Synonym(s):2,2'-((2R,2'R)-((4-(3-oxomorpholino)phenyl)azanediyl)bis(2-hydroxypropane-3,1-diyl))bis(isoindoline-1,3-dione)
- Description:Diphthalimido Morpholinone Analog
Specifications
About this item
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.