Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Digitalis, Reference Standard

Digitalis, Reference Standard

Digitalis, USP reference standard, 3 g
Katalogové číslo 1198000.
Dodavatel:   USP
Digitalis, Reference Standard
Katalogové číslo 1198000.
Dodavatel:  USP
Číslo CAS:  
Digitalis, Reference Standard

Digitalis, USP reference standard, 3 g

Číslo CAS:  
Price
6 720,00 Kč
Log in for availability
1 * 3 g
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

Specifications

Specifications

  • Pk:
    3 g
  • Standards:
    USP reference standard
  • Description:
    Digitalis
  • UN No.:
    1544
  • Cat. No.:
    1198000.
  • Description:
    Digitalis
  • CAS No.:
    8031-42-3

Specifications