Order Entry
Czech Republic
ContactUsLinkComponent
 
Desthiophenerivaroxaban Analog, Pharmaceutical Analytical Impurities
 :  USP
 
Desthiophenerivaroxaban Analog, Pharmaceutical Analytical Impurities
 :  USP

 

  • CAS No.:
    1424944-35-3
  • Synonym(s):
    (R)-2-((2-oxo-3-(4-(3-oxomorpholino)phenyl)oxazolidin-5-yl)methyl)isoindoline-1,3-dione
  • Description:
    Desthiophenerivaroxaban Phthalimodo Analog

 

Frequently Bought Together