Order Entry
Czech Republic
ContactUsLinkComponent
Budesonide D-homo Analog, Pharmaceutical Analytical Impurities
Budesonide D-homo Analog, Pharmaceutical Analytical Impurities
Catalog # 1A00500.
Supplier:  USP
CAS Number:  
Budesonide D-homo Analog, Pharmaceutical Analytical Impurities
Catalog # 1A00500.
Supplier:  USP
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    10 mg
  • Standards:
    Pharmaceutical Analytical Impurities
  • CAS No.:
    1040085-99-1
  • Description:
    Budesonide D-homo Analog
  • Synonym(s):
    (4aR,4bS,5S,6aS,7aR,10aR,11aS,11bS)-5-hydroxy-7a-(hydroxymethyl)-4a,6a-dimethyl-9-propyl-4b,6,6a,7a,10a,11,11a,11b,12,13-decahydrochryseno[2,3-d][1,3]dioxole-2,7(4aH,5H)-dione
  • Description:
    Budesonide D-homo Analog

Specifications

About this item

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.