Order Entry
Czech Republic
ContactUsLinkComponent
Bis(2-methylaminoethyl) Ether, Pharmaceutical Analytical Impurities
Bis(2-methylaminoethyl) Ether, Pharmaceutical Analytical Impurities
Catalog # 1A01350.
Supplier:  USP
CAS Number:  
Bis(2-methylaminoethyl) Ether, Pharmaceutical Analytical Impurities
Catalog # 1A01350.
Supplier:  USP
CAS Number:  

Specifications

  • Pack type:
    Ampule
  • Pk:
    25 mg
  • Standards:
    Pharmaceutical Analytical Impurities
  • CAS No.:
    3033-62-3
  • Description:
    Bis(2-methylaminoethyl) Ether
  • UN No.:
    2922
  • Synonym(s):
    2,2'-oxybis(N,N-dimethylethan-1-amine)
  • Description:
    Bis(2-methylaminoethyl) Ether

Specifications

About this item

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.