Alclometasone Dipropionate, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
2 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1012757.
- 1012746.
- Description:
- Alclometasone Dipropionate
- Alclometasone Dipropionate Related Compound A
- Synonym(s):
- 11β,17,21-trihydroxy-16α-methylpregna-1,4-diene-3,20-dione 17,21-dipropionate
- CAS No.:
- 66734-13-2
- Formula:
- C₂₈H₃₇ClO₇
Specifications
Product Family Options
Product Information
- StandardsPkAvailabilityPrice
- 1012757.Alclometasone DipropionateAlclometasone DipropionateUSP reference standard300 mgLog in for availability1 * 300 mg9 210,00 KčSpecifications:
More
Specifications
Cat. No.1012757.DescriptionAlclometasone DipropionateSynonym(s)CAS No.66734-13-2FormulaC₂₈H₃₇ClO₇ - 1012746.Alclometasone Dipropionate Related Compound AAlclometasone Dipropionate Related Compound AUSP reference standard20 mgLog in for availability1 * 20 mg12 200,00 KčSpecifications:
More
Specifications
Cat. No.1012746.DescriptionAlclometasone Dipropionate Related Compound ASynonym(s)11β,17,21-trihydroxy-16α-methylpregna-1,4-diene-3,20-dione 17,21-dipropionateCAS No.Formula



