Order Entry
Czech Republic
ContactUsLinkComponent
Abacavir Sulfate, Reference Standard
Abacavir Sulfate, Reference Standard
Supplier:  USP
Certificates
Abacavir Sulfate, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    188062-50-2
  • Storage conditions:
    Ambient
  • Description:
    Abacavir Sulfate
  • Synonym(s):
    4-[2-amino-6-(cyclopropylamino)-9H-purin-9yl]-2-cyclopentene-1-methanol sulfate (2:1)

Specifications

Frequently Bought Together