Order Entry
Czech Republic
ContactUsLinkComponent
6-Beta-Hydroxybudesonide, Pharmaceutical Analytical Impurities
6-Beta-Hydroxybudesonide, Pharmaceutical Analytical Impurities
Catalog # 1A01400.
Supplier:  USP
CAS Number:  
6-Beta-Hydroxybudesonide, Pharmaceutical Analytical Impurities
Catalog # 1A01400.
Supplier:  USP
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    25 mg
  • Standards:
    Pharmaceutical Analytical Impurities
  • CAS No.:
    88411-77-2
  • Description:
    6Beta-Hydroxybudesonide
  • Synonym(s):
    (2R,6aR,6bS,7S,8aS,8bS,11aR,12aS,12bS)-2,7-dihydroxy-8b-(2-hydroxyacetyl)-6a,8a-dimethyl-10-propyl-1,2,6a,6b,7,8,8a,8b,11a,12,12a,12b-dodecahydro-4H-naphtho[2',1':4,5]indeno[1,2-d][1,3]dioxol-4-one
  • Description:
    6Beta-Hydroxybudesonide

Specifications

About this item

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.