ContactUsLinkComponent
Sash-glider kit
Sash-glider kit
Catalog # LBCP3408500
Supplier:  LABCONCO
undefined
Sash-glider kit
Catalog # LBCP3408500
Supplier:  LABCONCO
Specifications
  • Description:
    Sash-glider kit
  • Pk:
    1 KIT
Specifications
About this item
Use the Sash-glider to clean both sides of a biological safety cabinet's sash while it remains closed, to avoid contamination risks.
Delivery: Sash-glider kit includes magnetic outside and inside blades, 2 microfibre sheets and 10 disposable white scrubbing strips. Additional scrubbing strips are available.