Order Entry
Czech Republic
ContactUsLinkComponent
DPDPB (1,4-Di[3'-(2'-pyridyldithio)propionamido]butane)
DPDPB (1,4-Di[3'-(2'-pyridyldithio)propionamido]butane)
Catalog # BC03
Supplier:  G-Biosciences
CAS Number:  
undefined
DPDPB (1,4-Di[3'-(2'-pyridyldithio)propionamido]butane)
Catalog # BC03
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Spacer Arm (Å):
    19.9
  • Reactive Toward:
    Sulfhydryls
  • Membrane Permeable:
    not done
  • Water Soluble:
    No
  • Cleavable/ Reversible:
    Reducing Agents (Thiols)

Specification Test Results

About this item

 :
 :482.71 g/mol
CAS Number:141647-62-3
Formula:C₂₀H₂₆N₄O₂S₄
MDL Number:MFCD00153604