Vertical-tube rotor package, VTi 50, Optima
521-3992
- Description:VTi 50 vertical tube rotor package. Vertical angle rotors feature the shortest pathlength of any rotor type, the fastest run time, and the highest resolution
- Pk:1
- For:Vertical angle rotor. A vertical rotor is used for: Isopycnic separations – specifically for the banding of DNA in cesium chloride. Gradient and density cushion separations, such as for viral particles. A gradient density cushion separation: Increasing density layers ending with a higher density cushion at the tube bottom. Ideal for isolating lagile, pelleted samples at the 'softer' cushion interface
- Max. RPM:50000
- No. of tubes accepted:8×39 ml, (1×3½"), 25×89 mm
A vertical rotor is used for: Isopycnic separations – specifically for the banding of DNA in cesium chloride. Gradient and density cushion separations, such as for viral particles. A gradient density cushion separation: Increasing density layers ending with a higher density cushion at the tube bottom. Ideal for isolating lagile, pelleted samples at the 'softer' cushion interface.