ContactUsLinkComponent
Fixed-angle titanium rotor package, Type 42.2 Ti, Optima
Fixed-angle titanium rotor package, Type 42.2 Ti, Optima
Catalog # 521-3750
Supplier:  Beckman Coulter
undefined
Fixed-angle titanium rotor package, Type 42.2 Ti, Optima
Catalog # 521-3750
Supplier:  Beckman Coulter
Specifications
  • Description:
    Type 42.2 Ti fixed angle titanium rotor package
  • Pk:
    1
  • For:
    Fixed angle (ultracentrifugation)
  • Max. RPM:
    42000
  • Radius:
    113 mm