ContactUsLinkComponent
Zonal Rotor with Standard Core, Titanium, Ti 15, Optima
Zonal Rotor with Standard Core, Titanium, Ti 15, Optima
Catalog # 521-4164
Supplier:  Beckman Coulter
Zonal Rotor with Standard Core, Titanium, Ti 15, Optima
Catalog # 521-4164
Supplier:  Beckman Coulter
Specifications
  • Description:
    Ti-15 Zonal rotor with standard core, titanium
  • Pk:
    1
  • For:
    Swinging bucket (High Performance)
  • Max. RPM:
    32000
  • Radius:
    88,9 mm