Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cesium carbonate
Cesium carbonate ≥99%, GPR RECTAPUR®
Cesium carbonate ≥99%, GPR RECTAPUR®
:
VWR Chemicals
Cesium carbonate ≥99%, GPR RECTAPUR®
:
VWR Chemicals
:
534-17-8
:
:
Cs₂CO₃
:
MFCD00010957
:
325.82 g/mol
:
13,02020
:
610 °C
:
4.24 g/cm³ (20 °C)
:
208-591-9
:
GHS05
:
GHS07
:
Ambient
Assay:
Min. 99 %
sds_VWRC91238
spec_VWRC91238.100
spec_VWRC91238.180
Frequently Bought Together
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Petroleum spirit, 40…60 °C, AnalaR NORMAPUR® ACS analytical reagent (max. 0.01% aromatic hydrocarbons)
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
: Merck
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals